missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetic acid, 99.5%, Thermo Scientific™

Catalog No. p-3758310
Change view
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126010050 5g Glass bottle
AC126010250 25g Glass bottle
AC126011000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC126010050 Supplier Thermo Scientific Chemicals Supplier No. 126010050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1798-09-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004334
InChI Key LEGPZHPSIPPYIO-UHFFFAOYSA-N
Synonym 3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my
PubChem CID 15719
IUPAC Name 2-(3-methoxyphenyl)acetic acid
SMILES COC1=CC=CC(=C1)CC(=O)O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99.50%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CO2H
Quantity 5g
Beilstein 10, I, 82
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 166.18
Percent Purity 99.5%
Chemical Name or Material 3-Methoxyphenylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 217-282-8

RUO – Research Use Only