Learn More
(3-Methoxyphenyl)acetonitrile, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 19924-43-7 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.18 |
| MDL Number | MFCD00001910 |
| InChI Key | LXKNAUOWEJWGTE-UHFFFAOYSA-N |
| Synonym | 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile |
| PubChem CID | 88310 |
| IUPAC Name | 2-(3-methoxyphenyl)acetonitrile |
| SMILES | COC1=CC=CC(=C1)CC#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC197870050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 19924-43-7 | |
| 147.18 | |
| LXKNAUOWEJWGTE-UHFFFAOYSA-N | |
| 88310 | |
| COC1=CC=CC(=C1)CC#N |
| C9H9NO | |
| MFCD00001910 | |
| 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile | |
| 2-(3-methoxyphenyl)acetonitrile |
Specifications
| 19924-43-7 | |
| 100.0 | |
| 164°C to 165°C (20.0mmHg) | |
| Authentic | |
| C9H9NO | |
| CH3OC6H4CH2CN | |
| 10, III, 429 | |
| 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile | |
| LXKNAUOWEJWGTE-UHFFFAOYSA-N | |
| 2-(3-methoxyphenyl)acetonitrile | |
| 88310 | |
| 97% |
| 96.0 | |
| 1.0500g/mL | |
| 98°C | |
| 96% min. (GC) | |
| 1.5297 to 1.5317 | |
| MFCD00001910 | |
| 1.05 | |
| Solubility in water: insoluble. Other solubilities: soluble in toluene | |
| COC1=CC=CC(=C1)CC#N | |
| 147.18 | |
| 147.18 | |
| (3-Methoxyphenyl)acetonitrile, 97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
GHS Signal Word: Danger
EINECSNumber : 243-428-5
TSCA : TSCA
RUO – Research Use Only