missing translation for 'onlineSavingsMsg'
Learn More

3-(Methoxydimethylsilyl)propyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. M25255G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25255G Supplier TCI America Supplier No. M25255G

Chemical Identifiers

CAS 111918-90-2
Molecular Formula C9H18O3Si
Molecular Weight (g/mol) 202.325
MDL Number MFCD00054801
InChI Key ZCRUJAKCJLCJCP-UHFFFAOYSA-N
Synonym Acrylic Acid 3-(Methoxydimethylsilyl)propyl Ester
PubChem CID 9813088
IUPAC Name 3-[methoxy(dimethyl)silyl]propyl prop-2-enoate
SMILES CO[Si](C)(C)CCCOC(=O)C=C

Specifications

Color Colorless
Quantity 5 g
Formula Weight 202.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Methoxydimethylsilyl)propyl Acrylate (stabilized with MEHQ)
TSCA No
Recommended Storage Freezer