missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxycinnamic acid, predominantly trans, 98+%

Catalog No. p-7080958
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1072306 5 g
AAA1072314 25 g
AAA1072322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1072306 Supplier Thermo Scientific Chemicals Supplier No. A1072306
Only null left

CAS: 1533748 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1533748
MDL Number MFCD00004386
ChEBI CHEBI:64746

Specifications

Melting Point 117°C to 121°C
Molecular Formula C6H7K3O8
Quantity 5 g
Beilstein 2209732
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
Molecular Weight (g/mol) 324.41
PubChem CID 2735208
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material 3-Methoxycinnamic acid, predominantly trans
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 228-049-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only