missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxycarbonylphenylboronic acid, 97%

Catalog No. p-3736852
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369830010 1 g Glass bottle
AC369830050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC369830010 Supplier Thermo Scientific Chemicals Supplier No. 369830010
Only null left

CAS: 99769-19-4 | C8H9BO4 | 179.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99769-19-4
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD02093046
InChI Key ALTLCJHSJMGSLT-UHFFFAOYSA-N
Synonym 3-methoxycarbonyl phenylboronic acid, 3-methoxycarbonylphenyl boronic acid, 3-methoxycarbonyl phenyl boronic acid, 3-methoxycarbonyl benzeneboronic acid, 3-methoxycarbonylphenylbaronic acid, methyl 3-dihydroxyboranyl benzoate, m-methoxycarbonyl phenylboronic acid, 3-carbomethoxy-phenylboronic acid
PubChem CID 2734714
IUPAC Name (3-methoxycarbonylphenyl)boronic acid
SMILES COC(=O)C1=CC=CC(=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 187°C to 191°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 179.97
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Methoxycarbonylphenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only