missing translation for 'onlineSavingsMsg'
Learn More

3-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M19061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19061G Supplier TCI America Supplier No. M19061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99769-19-4
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD02093046
InChI Key ALTLCJHSJMGSLT-UHFFFAOYSA-N
Synonym 3-methoxycarbonyl phenylboronic acid, 3-methoxycarbonylphenyl boronic acid, 3-methoxycarbonyl phenyl boronic acid, 3-methoxycarbonyl benzeneboronic acid, 3-methoxycarbonylphenylbaronic acid, methyl 3-dihydroxyboranyl benzoate, m-methoxycarbonyl phenylboronic acid, 3-carbomethoxy-phenylboronic acid
PubChem CID 2734714
IUPAC Name [3-(methoxycarbonyl)phenyl]boronic acid
SMILES COC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 209°C
Color White-Yellow
Quantity 1 g
Formula Weight 179.97
Physical Form Crystalline Powder
Chemical Name or Material 3-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)