missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7133524
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29925G 5 g
M299225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29925G Supplier TCI America Supplier No. M29925G
Only null left

Chemical Identifiers

CAS 1711-05-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000673
InChI Key RUQIUASLAXJZIE-UHFFFAOYSA-N
Synonym m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid
PubChem CID 74374
IUPAC Name 3-methoxybenzoyl chloride
SMILES COC1=CC=CC(=C1)C(=O)Cl

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
UN Number 1729
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzoyl Chloride