missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzoyl chloride, 99%

Catalog No. p-7027091
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2357109 10 g
AAB2357118 50 g
AAB2357130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2357109 Supplier Thermo Scientific Chemicals Supplier No. B2357109

CAS: 1711-05-3 | C8H7ClO2 | 170.592 g/mol

3-Methoxybenzoyl chloride is used in the synthesis of 2-arylbenzofuran-based molecules that act against Alzheimer’s disease, combating symptoms. In addition, it aids in the optimization of dihydropyrrolopyrimidine inhibitors against PI3K (phosphoinositide 3-kinase) resulting in tumor growth inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform. Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.

Chemical Identifiers

CAS 1711-05-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000673
InChI Key RUQIUASLAXJZIE-UHFFFAOYSA-N
Synonym m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid
PubChem CID 74374
IUPAC Name 3-methoxybenzoyl chloride
SMILES COC1=CC=CC(=C1)C(=O)Cl

Specifications

Density 1.214
Boiling Point 123°C to 125°C (15 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.5586
Quantity 10 g
UN Number UN1729
Beilstein 386659
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform. Reacts with water.
Formula Weight 170.6
Percent Purity 99%
Chemical Name or Material 3-Methoxybenzoyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANISOYL CHLORIDE
EINECSNumber 216-975-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only