missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzohydrazide 98.0+%, TCI America™

Catalog No. p-7117441
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14465G 5 g
A144625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14465G Supplier TCI America Supplier No. A14465G

Chemical Identifiers

CAS 5785-06-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00007601
InChI Key VMZSDAQEWPNOIB-UHFFFAOYSA-N
Synonym 3-methoxybenzhydrazide, m-anisic hydrazide, m-anisohydrazide, m-methoxy benzhydrazide, 3-methoxybenzoic hydrazide, benzoylhydrazine, m-methoxy, 3-methoxybenzenecarbohydrazide, 3-methoxybenzene-1-carbohydrazide, benzoic acid, 3-methoxy-, hydrazide, m-anisoylhydrazine
PubChem CID 79848
IUPAC Name 3-methoxybenzohydrazide
SMILES COC1=CC=CC(=C1)C(=O)NN

Specifications

Melting Point 93°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxybenzohydrazide