missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzenesulfonyl chloride, 97%

Catalog No. p-7058470
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1705003 1 g
AAL1705006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1705003 Supplier Thermo Scientific Chemicals Supplier No. L1705003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10130-74-2 | C7H7ClO3S | 206.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10130-74-2
Molecular Formula C7H7ClO3S
Molecular Weight (g/mol) 206.64
MDL Number MFCD01318155
InChI Key JHJKSEKUZNJKGO-UHFFFAOYSA-N
Synonym 3-methoxybenzene-1-sulfonyl chloride, 3-methoxy-benzenesulfonyl chloride, 3-methoxybenzenesulphonyl chloride, m-anisolesulfonyl chloride, 3-methoxybenzenesulfonylchloride, benzenesulfonyl chloride, 3-methoxy, akos bb-9459, 3-methoxyphenylsulfonyl chloride, 3-methoxybenzene sulfonyl chloride, m-anisolesulphonyl chloride
PubChem CID 2737504
IUPAC Name 3-methoxybenzenesulfonyl chloride
SMILES COC1=CC(=CC=C1)S(=O)(=O)Cl

Specifications

Density 1.46
Boiling Point 158°C to 159°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.557
Quantity 1 g
UN Number UN3265
Beilstein 2093871
Sensitivity Moisture sensitive
Formula Weight 206.65
Percent Purity 97%
Chemical Name or Material 3-Methoxybenzenesulfonyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only