missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyazetidine hydrochloride, 95%

Catalog No. AAH57769MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57769MD 250 mg
AAH57769ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57769MD Supplier Thermo Scientific Chemicals Supplier No. H57769MD

CAS: 148644-09-1 | C4H10ClNO | 123.58 g/mol

It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.

Chemical Identifiers

CAS 148644-09-1
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD06804514
InChI Key KSXGQRBTBLQJEF-UHFFFAOYSA-N
Synonym 3-methoxyazetidine hydrochloride, 3-methoxy-azetidine hydrochloride, 3-azetidinyl methyl ether hydrochloride, 3-methoxyazetidine hcl, azetidine, 3-methoxy-, hydrochloride, azetidine, 3-methoxy-,hydrochloride 1:1, acmc-20a0d9, ksc495e1t, 3-methoxyazetidinehydrochloride, 3-methoxy-azetidine-hydrochloride
PubChem CID 22242858
IUPAC Name 3-methoxyazetidine;hydrochloride
SMILES COC1CNC1.Cl

Specifications

Boiling Point 143°C to 149°C
Quantity 250 mg
Formula Weight 123.58
Percent Purity 95%
Chemical Name or Material 3-Methoxyazetidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only