missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-nitrobenzaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 53055-05-3
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007135
InChI Key GDTUACILWWLIJF-UHFFFAOYSA-N
Synonym benzaldehyde, 3-methoxy-2-nitro, 3-methoxy-2-nitro-benzaldehyde, 2-nitro-3-methoxybenzaldehyde, pubchem13093, acmc-20agz4, 2-nitro-3-methoxy benzaldehyde, benzaldehyde,3-methoxy-2-nitro, 3-methoxy-2-nitrobenzaldehyde
PubChem CID 98867
IUPAC Name 3-methoxy-2-nitrobenzaldehyde
SMILES COC1=CC=CC(C=O)=C1[N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC172670010
SDS View Documents
Thermo Scientific Chemicals
172670010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC172670050
SDS View Documents
Thermo Scientific Chemicals
172670050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC172670250
SDS View Documents
Thermo Scientific Chemicals
172670250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

53055-05-3
181.15
GDTUACILWWLIJF-UHFFFAOYSA-N
98867
COC1=CC=CC(C=O)=C1[N+]([O-])=O
C8H7NO4
MFCD00007135
benzaldehyde, 3-methoxy-2-nitro, 3-methoxy-2-nitro-benzaldehyde, 2-nitro-3-methoxybenzaldehyde, pubchem13093, acmc-20agz4, 2-nitro-3-methoxy benzaldehyde, benzaldehyde,3-methoxy-2-nitro, 3-methoxy-2-nitrobenzaldehyde
3-methoxy-2-nitrobenzaldehyde

Specifications

53055-05-3
100.0
96% min. (GC)
C8H7NO4
MFCD00007135
08, 62
GDTUACILWWLIJF-UHFFFAOYSA-N
3-methoxy-2-nitrobenzaldehyde
98867
97%
96.0
Authentic
Glass bottle
CH3OC6H3(NO2)CHO
1g
benzaldehyde, 3-methoxy-2-nitro, 3-methoxy-2-nitro-benzaldehyde, 2-nitro-3-methoxybenzaldehyde, pubchem13093, acmc-20agz4, 2-nitro-3-methoxy benzaldehyde, benzaldehyde,3-methoxy-2-nitro, 3-methoxy-2-nitrobenzaldehyde
COC1=CC=CC(C=O)=C1[N+]([O-])=O
181.15
181.15
3-Methoxy-2-nitrobenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-332-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only