missing translation for 'onlineSavingsMsg'
Learn More

3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane, 98%

Catalog No. AAL1668214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1668214 25 g
AAL1668222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1668214 Supplier Thermo Scientific Chemicals Supplier No. L1668214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17096-07-0 | C16H38O5Si4 | 422.815 g/mol

3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agent.

Chemical Identifiers

CAS 17096-07-0
Molecular Formula C16H38O5Si4
Molecular Weight (g/mol) 422.815
MDL Number MFCD00053871
InChI Key BESKSSIEODQWBP-UHFFFAOYSA-N
Synonym 3-methacryloyloxy propyltris trimethylsiloxy silane, 3-tris trimethylsiloxy silyl propyl methacrylate, 3-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxan-3-yl propyl methacrylate, unii-d32s78j7t8, 3-methacryloyloxypropyl tris trimethylsiloxy silane, 3-methacryloxypropyltris trimethylsiloxy silane, 3-tris trimethylsilyloxy silyl propyl methacrylate, methacryloxypropyltris trimethylsiloxy silane, 3-3,3,3-trimethyl-1,1-bis trimethylsilyl oxy disiloxanyl propyl methacrylate
PubChem CID 123371
IUPAC Name 3-tris(trimethylsilyloxy)silylpropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 112°C to 115°C (0.2mmHg)
Quantity 25 g
Flash Point 201°C (393°F)
Solubility Information Immiscible with water.
Formula Weight 422.82
Percent Purity 98%
Refractive Index 1.4175
Density 0.93
Chemical Name or Material 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-165-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only