missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methacrylamidophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$156.59 - $156.59
Specifications
| CAS | 48150-45-4 |
|---|---|
| Color | Yellow |
| Molecular Formula | C10H12BNO3 |
| MDL Number | MFCD09475805 |
| Synonym | 3-Methacrylamidobenzeneboronic Acid |
Specifications
| 48150-45-4 | |
| C10H12BNO3 | |
| 3-Methacrylamidobenzeneboronic Acid | |
| B(C1=CC(=CC=C1)NC(=O)C(=C)C)(O)O | |
| 205.02 | |
| 205.02 |
| Yellow | |
| MFCD09475805 | |
| GBBUBIKYAQLESK-UHFFFAOYSA-N | |
| [3-(2-methylprop-2-enoylamino)phenyl]boronic acid | |
| 3016474 | |
| Crystalline Powder |