missing translation for 'onlineSavingsMsg'
Learn More

3-Methacrylamidophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M26761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M26761G Supplier TCI America Supplier No. M26761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Methacrylamidophenylboronic Acid (contains varying amounts of Anhydride)
CAS 48150-45-4
Color Yellow
Molecular Formula C10H12BNO3
Quantity 1 g
MDL Number MFCD09475805
Synonym 3-Methacrylamidobenzeneboronic Acid
InChI Key GBBUBIKYAQLESK-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)NC(=O)C(=C)C)(O)O
IUPAC Name [3-(2-methylprop-2-enoylamino)phenyl]boronic acid
Molecular Weight (g/mol) 205.02
PubChem CID 3016474
Formula Weight 205.02
Physical Form Crystalline Powder
Show More Show Less