missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)triethoxysilane 96.0+%, TCI America™

Catalog No. p-7132764
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
M150525G 25 g
M1505100G 100 g
M1505500G 500 g
3 options

Catalog No. M150525G

Supplier: TCI America M150525G

Only null left

Chemical Identifiers

CAS 14814-09-6
Molecular Formula C9H22O3SSi
Molecular Weight (g/mol) 238.417
MDL Number MFCD00053924
InChI Key DCQBZYNUSLHVJC-UHFFFAOYSA-N
Synonym 3-mercaptopropyltriethoxysilane, 3-mercaptopropyl triethoxysilane, 3-triethoxysilyl propane-1-thiol, 1-propanethiol, 3-triethoxysilyl, 3-triethoxysilyl propanethiol, unii-z069rx894n, triethoxy 3-mercaptopropyl silane, 3-triethoxysilyl propyl mercaptan, gamma-mercaptopropyl triethoxysilane, silane, 3-mercaptopropyl triethoxy
PubChem CID 84658
IUPAC Name 3-triethoxysilylpropane-1-thiol
SMILES CCO[Si](CCCS)(OCC)OCC

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 238.42
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Mercaptopropyl)triethoxysilane
EINECSNumber (2)-2045
RTECSNumber TZ7760000
TSCA Yes