missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)triethoxysilane, 94%

Catalog No. AAB2119122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2119122 100 g
AAB2119114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2119122 Supplier Thermo Scientific Chemicals Supplier No. B2119122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14814-09-6 | C9H22O3SSi | 238.417 g/mol

It can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.

Solubility
Hydrolyzes slowly in water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 14814-09-6
Molecular Formula C9H22O3SSi
Molecular Weight (g/mol) 238.417
MDL Number MFCD00053924
InChI Key DCQBZYNUSLHVJC-UHFFFAOYSA-N
Synonym 3-mercaptopropyltriethoxysilane, 3-mercaptopropyl triethoxysilane, 3-triethoxysilyl propane-1-thiol, 1-propanethiol, 3-triethoxysilyl, 3-triethoxysilyl propanethiol, unii-z069rx894n, triethoxy 3-mercaptopropyl silane, 3-triethoxysilyl propyl mercaptan, gamma-mercaptopropyl triethoxysilane, silane, 3-mercaptopropyl triethoxy
PubChem CID 84658
IUPAC Name 3-triethoxysilylpropane-1-thiol
SMILES CCO[Si](CCCS)(OCC)OCC

Specifications

Boiling Point 210°C
Quantity 100 g
Linear Formula (CH3CH2O)3Si(CH2)3SH
Beilstein 2039575
Flash Point 88°C (190°F)
Solubility Information Hydrolyzes slowly in water.
Formula Weight 238.42
Percent Purity 94%
Odor Unpleasant
Refractive Index 1.437
Sensitivity Air and moisture sensitive
Density 0.98
Chemical Name or Material (3-Mercaptopropyl)triethoxysilane
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 238-883-1
RTECSNumber TZ7760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only