missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)methyldimethoxysilane, 95%

Catalog No. AA42426AK
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42426AK 250 mL
AA42426AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42426AK Supplier Thermo Scientific Chemicals Supplier No. 042426AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31001-77-1 | C6H16O2SSi | 180.337 g/mol

(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31001-77-1
Molecular Formula C6H16O2SSi
Molecular Weight (g/mol) 180.337
MDL Number MFCD00053923
InChI Key IKYAJDOSWUATPI-UHFFFAOYSA-N
Synonym 3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan
PubChem CID 2759524
IUPAC Name 3-[dimethoxy(methyl)silyl]propane-1-thiol
SMILES CO[Si](C)(CCCS)OC

Specifications

Boiling Point 96°C (30mmHg)
Color Colorless
Physical Form Liquid
Quantity 250 mL
Flash Point 93°C (199°F)
Refractive Index 1.45
Sensitivity Moisture sensitive
Density 1.000 g/mL
Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
Solubility Information Insoluble in water.
Formula Weight 180.34
Percent Purity 95%
Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane
Show More Show Less
Hazard Category H227-H302-H317
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c
EINECSNumber 250-426-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only