missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopropionic Acid 98.0+%, TCI America™

Catalog No. m0061100g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
M0061100G 100 g
M006125G 25 g
M0061500G 500 g
3 options

Catalog No. M0061100G

Supplier: TCI America M0061100G

Only null left

Chemical Identifiers

CAS 107-96-0
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00004897
InChI Key DKIDEFUBRARXTE-UHFFFAOYSA-N
Synonym 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid
PubChem CID 6514
ChEBI CHEBI:44111
IUPAC Name 3-sulfanylpropanoic acid
SMILES C(CS)C(=O)O

Specifications

Color Colorless
Boiling Point 116°C
Quantity 100 g
UN Number 2922
Formula Weight 106.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Mercaptopropionic Acid
EINECSNumber (2)-1381
RTECSNumber UF5270000
TSCA Yes