missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopropionic Acid 98.0+%, TCI America™

Catalog No. M006125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M006125G 25 g
M0061100G 100 g
M0061500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M006125G Supplier TCI America Supplier No. M006125G

Chemical Identifiers

CAS 107-96-0
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00004897
InChI Key DKIDEFUBRARXTE-UHFFFAOYSA-N
Synonym 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid
PubChem CID 6514
ChEBI CHEBI:44111
IUPAC Name 3-sulfanylpropanoic acid
SMILES C(CS)C(=O)O

Specifications

Color Colorless
Boiling Point 116°C
Quantity 25 g
UN Number 2922
Formula Weight 106.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Mercaptopropionic Acid
EINECSNumber (2)-1381
RTECSNumber UF5270000
TSCA Yes