Learn More
3-Mercaptopropionic acid, 99%
CAS: 107-96-0 | C3H6O2S | 106.139 g/mol
Supplier: Thermo Scientific Chemicals A1326136
Description
3-Mercaptopropionic acid is widely used in food and beverage industries as a flavoring agent. It is used in the production of PVC stabilizers, which are used as chain transfer agents in polymerizations. It can be used as primary or secondary, color stabilizer in combination with phenolic antioxidant for polymers. It acts as a sulfide ion equivalent and is utilized in the preparation of diaryl sulfide from aryl iodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Mercaptopropionic acid is widely used in food and beverage industries as a flavoring agent. It is used in the production of PVC stabilizers, which are used as chain transfer agents in polymerizations. It can be used as primary or secondary, color stabilizer in combination with phenolic antioxidant for polymers. It acts as a sulfide ion equivalent and is utilized in the preparation of diaryl sulfide from aryl iodide.
Solubility
Miscible with water, alcohol, benzene and ether.
Notes
Air Sensitive. Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.
Specifications
| 3-Mercaptopropionic acid | |
| 17°C to 19°C | |
| 110°C to 111°C (15 mmHg) | |
| C3H6O2S | |
| HSCH2CH2CO2H | |
| 500 g | |
| 773807 | |
| Miscible with water,alcohol,benzene and ether. | |
| C(CS)C(=O)O | |
| 106.139 | |
| CHEBI:44111 | |
| 99% |
| 107-96-0 | |
| 1.22 | |
| 93°C (199°F) | |
| 1.492 | |
| MFCD00004897 | |
| UN2922 | |
| 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid | |
| DKIDEFUBRARXTE-UHFFFAOYSA-N | |
| 3-sulfanylpropanoic acid | |
| 6514 | |
| 106.14 |
Chemical Identifiers
| 107-96-0 | |
| 106.139 | |
| DKIDEFUBRARXTE-UHFFFAOYSA-N | |
| 6514 | |
| 3-sulfanylpropanoic acid |
| C3H6O2S | |
| MFCD00004897 | |
| 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid | |
| CHEBI:44111 | |
| C(CS)C(=O)O |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P234-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
H227-H290-H301-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 203-537-0
RTECSNumber : UF5270000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only