missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopropionic acid, 99%

Catalog No. AAA1326122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1326122 100 g
AAA1326136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1326122 Supplier Thermo Scientific Chemicals Supplier No. A1326122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-96-0 | C3H6O2S | 106.139 g/mol

3-Mercaptopropionic acid is widely used in food and beverage industries as a flavoring agent. It is used in the production of PVC stabilizers, which are used as chain transfer agents in polymerizations. It can be used as primary or secondary, color stabilizer in combination with phenolic antioxidant for polymers. It acts as a sulfide ion equivalent and is utilized in the preparation of diaryl sulfide from aryl iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Mercaptopropionic acid is widely used in food and beverage industries as a flavoring agent. It is used in the production of PVC stabilizers, which are used as chain transfer agents in polymerizations. It can be used as primary or secondary, color stabilizer in combination with phenolic antioxidant for polymers. It acts as a sulfide ion equivalent and is utilized in the preparation of diaryl sulfide from aryl iodide.

Solubility
Miscible with water, alcohol, benzene and ether.

Notes
Air Sensitive. Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 107-96-0
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00004897
InChI Key DKIDEFUBRARXTE-UHFFFAOYSA-N
Synonym 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid
PubChem CID 6514
ChEBI CHEBI:44111
IUPAC Name 3-sulfanylpropanoic acid
SMILES C(CS)C(=O)O

Specifications

Melting Point 17°C to 19°C
Density 1.22
Boiling Point 110°C to 111°C (15 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.492
Linear Formula HSCH2CH2CO2H
Quantity 100 g
UN Number UN2922
Beilstein 773807
Solubility Information Miscible with water,alcohol,benzene and ether.
Formula Weight 106.14
Percent Purity 99%
Chemical Name or Material 3-Mercaptopropionic acid
Show More Show Less
Hazard Category H227-H290-H301-H314-H332-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P234-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-537-0
RTECSNumber UF5270000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only