Learn More
3-Mercaptopropionic acid, 98%
CAS: 107-96-0 | C3H6O2S | 106.14 g/mol
Supplier: Thermo Scientific Chemicals 125535000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| 3-Mercaptopropionic acid | |
| 99.0 | |
| 17.0°C to 19.0°C | |
| 1.2180g/mL | |
| 124°C | |
| 99.0 to 101% (Iodimetry) | |
| C3H6O2S | |
| HSCH2CH2CO2H | |
| 500 mL | |
| 1.218 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol | |
| C(CS)C(=O)O | |
| 106.14 | |
| CHEBI:44111 | |
| 99% |
| 107-96-0 | |
| 100.0 | |
| White | |
| 110.0°C to 111.0°C (15.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4910 to 1.4930 | |
| MFCD00004897 | |
| 03,299 | |
| 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid | |
| DKIDEFUBRARXTE-UHFFFAOYSA-N | |
| 3-sulfanylpropanoic acid | |
| 6514 | |
| 106.14 | |
| Crystalline Powder, Crystals or Chunks |
Chemical Identifiers
| 107-96-0 | |
| 106.14 | |
| DKIDEFUBRARXTE-UHFFFAOYSA-N | |
| 6514 | |
| 3-sulfanylpropanoic acid |
| C3H6O2S | |
| MFCD00004897 | |
| 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid | |
| CHEBI:44111 | |
| C(CS)C(=O)O |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 203-537-0
RTECSNumber : UF5270000
TSCA : TSCA
RUO – Research Use Only