missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopropionic acid, 98%

Catalog No. AC125531000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC125531000 100 mL
AC125535000 500 mL
AC125530025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125531000 Supplier Thermo Scientific Chemicals Supplier No. 125531000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-96-0 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 107-96-0
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004897
InChI Key DKIDEFUBRARXTE-UHFFFAOYSA-N
Synonym 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid
PubChem CID 6514
ChEBI CHEBI:44111
IUPAC Name 3-sulfanylpropanoic acid
SMILES C(CS)C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 17.0°C to 19.0°C
Color White
Density 1.2180g/mL
Boiling Point 110.0°C to 111.0°C (15.0 mmHg)
Flash Point 124°C
Infrared Spectrum Authentic
Assay Percent Range 99.0 to 101% (Iodimetry)
Packaging Glass bottle
Refractive Index 1.4910 to 1.4930
Linear Formula HSCH2CH2CO2H
Quantity 100 mL
Beilstein 03,299
Specific Gravity 1.218
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 106.14
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 3-Mercaptopropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 203-537-0
RTECSNumber UF5270000
TSCA TSCA

RUO – Research Use Only