missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-2-pentanone, 97%, stab. with 0.1% Calcium carbonate

Catalog No. AAB2179706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2179706 5 g
AAB2179703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2179706 Supplier Thermo Scientific Chemicals Supplier No. B2179706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 67633-97-0 | C5H10OS | 118.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67633-97-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD02093290
InChI Key SZECUQRKLXRGSJ-UHFFFAOYSA-N
Synonym 3-mercapto-2-pentanone, 2-pentanone, 3-mercapto, 3-mercaptopentan-2-one, fema no. 3300, 3-mercapto-2-penanone, 2-pentanone,3-mercapto, acmc-1b6ys, 2-pentanone, 3-mercapto-7ci,9ci
PubChem CID 62237
IUPAC Name 3-sulfanylpentan-2-one
SMILES CCC(C(=O)C)S

Specifications

Density 0.988
Refractive Index 1.466
Quantity 5 g
UN Number UN3336
Sensitivity Light sensitive
Formula Weight 118.19
Percent Purity 98%
Chemical Name or Material 3-Mercapto-2-pentanone, stabilized with 0.1% Calcium carbonate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
EINECSNumber 266-799-5
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only