missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-2-butanone, 98%, stab. with 0.1% Calcium carbonate

CAS: 40789-98-8 | C4H8OS | 104.17 g/mol

$379.53 - $379.53

Chemical Identifiers

CAS 40789-98-8
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.17
MDL Number MFCD00085206
InChI Key XLMPYCGSRHSSSX-UHFFFAOYNA-N
Synonym 3-mercapto-2-butanone, 2-butanone, 3-mercapto, 2-mercapto-3-butanone, 3-mercaptobutan-2-one, fema no. 3298, mercapto-3 butanone-2 french, mercapto-3 butanone-2, acmc-209jgq, 4-01-00-03993 beilstein handbook reference
PubChem CID 38679
IUPAC Name 3-sulfanylbutan-2-one
SMILES CC(S)C(C)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2249414
View Documents
Thermo Scientific Chemicals
B2249414
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40789-98-8
104.17
XLMPYCGSRHSSSX-UHFFFAOYNA-N
38679
CC(S)C(C)=O
C4H8OS
MFCD00085206
3-mercapto-2-butanone, 2-butanone, 3-mercapto, 2-mercapto-3-butanone, 3-mercaptobutan-2-one, fema no. 3298, mercapto-3 butanone-2 french, mercapto-3 butanone-2, acmc-209jgq, 4-01-00-03993 beilstein handbook reference
3-sulfanylbutan-2-one

Specifications

40789-98-8
50°C (18 mmHg)
C4H8OS
MFCD00085206
UN1224
3-mercapto-2-butanone, 2-butanone, 3-mercapto, 2-mercapto-3-butanone, 3-mercaptobutan-2-one, fema no. 3298, mercapto-3 butanone-2 french, mercapto-3 butanone-2, acmc-209jgq, 4-01-00-03993 beilstein handbook reference
CC(S)C(C)=O
104.17
104.17
3-Mercapto-2-butanone, stabilized with 0.1% Calcium carbonate
1.015
47°C (117°F)
1.475
25 g
Light sensitive
XLMPYCGSRHSSSX-UHFFFAOYNA-N
3-sulfanylbutan-2-one
38679
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 255-082-2

RTECSNumber : EL9050000

TSCA : Yes

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only