missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1,2,4-triazole 98.0+%, TCI America™

Catalog No. M081425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M081425G 25 g
M0814100G 100 g
M0814500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M081425G Supplier TCI America Supplier No. M081425G

Chemical Identifiers

CAS 3179-31-5
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.13
MDL Number MFCD00005229
InChI Key AFBBKYQYNPNMAT-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol
PubChem CID 2723802
IUPAC Name 2,3-dihydro-1H-1,2,4-triazole-3-thione
SMILES S=C1NNC=N1

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 25 g
Formula Weight 101.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Mercapto-1,2,4-triazole
EINECSNumber (5)-0606
RTECSNumber XZ5267500
TSCA Yes