Learn More
3-Mercapto-1,2,4-triazole, 98%
CAS: 3179-31-5 | C2H3N3S | 101.13 g/mol
Supplier: Thermo Scientific Chemicals A1312014
Description
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
Solubility
Soluble in hot water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 3-Mercapto-1,2,4-triazole | |
| 217°C to 224°C | |
| MFCD00005229 | |
| 107731 | |
| Soluble in hot water. | |
| S=C1NNC=N1 | |
| 101.13 | |
| 101.13 |
| 3179-31-5 | |
| C2H3N3S | |
| 25 g | |
| 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol | |
| AFBBKYQYNPNMAT-UHFFFAOYSA-N | |
| 1,2-dihydro-1,2,4-triazole-3-thione | |
| 2723802 | |
| 98% |
Chemical Identifiers
| 3179-31-5 | |
| 101.13 | |
| AFBBKYQYNPNMAT-UHFFFAOYSA-N | |
| 2723802 |
| C2H3N3S | |
| MFCD00005229 | |
| 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol | |
| S=C1NNC=N1 |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 221-656-6
RTECSNumber : XZ5267500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only