missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1,2,4-triazole, 98%

Catalog No. AAA1312006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1312006 5 g
AAA1312014 25 g
AAA1312022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1312006 Supplier Thermo Scientific Chemicals Supplier No. A1312006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3179-31-5 | C2H3N3S | 101.13 g/mol

3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.

Solubility
Soluble in hot water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3179-31-5
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.13
MDL Number MFCD00005229
InChI Key AFBBKYQYNPNMAT-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol
PubChem CID 2723802
SMILES S=C1NNC=N1

Specifications

Melting Point 217°C to 224°C
Quantity 5 g
Beilstein 107731
Solubility Information Soluble in hot water.
IUPAC Name 1,2-dihydro-1,2,4-triazole-3-thione
Formula Weight 101.13
Percent Purity 98%
Chemical Name or Material 3-Mercapto-1,2,4-triazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-656-6
RTECSNumber XZ5267500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only