missing translation for 'onlineSavingsMsg'
Learn More

3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%

Catalog No. p-7077410
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453251000 100 mg
AC453250010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453251000 Supplier Thermo Scientific Chemicals Supplier No. 453251000
Only null left

CAS: 55750-62-4 | C11H10N2O6 | 266.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55750-62-4
Molecular Formula C11H10N2O6
Molecular Weight (g/mol) 266.21
InChI Key JKHVDAUOODACDU-UHFFFAOYSA-N
Synonym n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester
PubChem CID 4620597
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O

Specifications

Melting Point 159.0°C to 160.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 100 mg
Formula Weight 266.21
Percent Purity 99%
Chemical Name or Material 3-Maleimidopropionic acid N-hydroxysuccinimide ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only