Learn More
Chemglass Inc 3000mL Glas-Col Heating Mantle, Series TM, Spherical, 500 Watts
Catalog No. CG1200010
Encompass
3000mL Glas-Col Heating Mantle, Series TM, Spherical, 500 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. Mantles are 115 Volt.
Catalog No. CG1200010 Supplier Chemglass Inc Supplier No. CG1200010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3000mL Glas-Col Heating Mantle, Series TM, Spherical, 500 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. Mantles are 115 Volt.