missing translation for 'onlineSavingsMsg'
Learn More

3-Isochromanone, Thermo Scientific Chemicals

Catalog No. AC370130010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC370130010 1 g
AC370130050 5 g
AC370130250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC370130010 Supplier Thermo Scientific Chemicals Supplier No. 370130010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4385-35-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
InChI Key ILHLUZUMRJQEAH-UHFFFAOYSA-N
IUPAC Name 3,4-dihydro-1H-2-benzopyran-3-one
SMILES O=C1CC2=CC=CC=C2CO1

Specifications

Melting Point 83.0°C to 86.0°C
Quantity 1 g
Beilstein 17, V,10, 19
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
Formula Weight 148.16
Chemical Name or Material 3-Isochromanone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 224-493-9

RUO – Research Use Only