missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isochromanone 99.0+%, TCI America™
Supplier: TCI America I054425G
Specifications
| 3-Isochromanone | |
| 83°C | |
| 130°C | |
| MFCD00043005 | |
| 1,4-Dihydro-3H-2-benzopyran-3-one | |
| C1C2=CC=CC=C2COC1=O | |
| 148.161 | |
| CHEBI:28170 | |
| ≥99.0% (GC) |
| 4385-35-7 | |
| White-Yellow | |
| C9H8O2 | |
| 25 g | |
| ILHLUZUMRJQEAH-UHFFFAOYSA-N | |
| 1,4-dihydroisochromen-3-one | |
| 78092 | |
| 148.16 | |
| Crystalline Powder |
Chemical Identifiers
| 4385-35-7 | |
| 148.161 | |
| ILHLUZUMRJQEAH-UHFFFAOYSA-N | |
| 78092 | |
| 1,4-dihydroisochromen-3-one |
| C9H8O2 | |
| MFCD00043005 | |
| 1,4-Dihydro-3H-2-benzopyran-3-one | |
| CHEBI:28170 | |
| C1C2=CC=CC=C2COC1=O |
Safety and Handling
EINECSNumber : (5)-6813
TSCA : No