missing translation for 'onlineSavingsMsg'
Learn More

3-Isochromanone 99.0+%, TCI America™

Catalog No. I05441G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05441G 1 g
I05445G 5 g
I054425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I05441G Supplier TCI America Supplier No. I05441G

Chemical Identifiers

CAS 4385-35-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00043005
InChI Key ILHLUZUMRJQEAH-UHFFFAOYSA-N
Synonym 1,4-Dihydro-3H-2-benzopyran-3-one
PubChem CID 78092
ChEBI CHEBI:28170
IUPAC Name 1,4-dihydroisochromen-3-one
SMILES C1C2=CC=CC=C2COC1=O

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 148.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Isochromanone
EINECSNumber (5)-6813
TSCA No