Learn More
Thermo Scientific Chemicals 3-Isobutyl-1-methylxanthine, 99+%
CAS: 28822-58-4 | C10H14N4O2 | 222.25 g/mol
Supplier: Thermo Scientific Chemicals 228420050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Isobutyl-1-methylxanthine | |
| 99.0 | |
| 200.0°C to 203.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005584 | |
| 26, IV, 2350 | |
| Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,2-propanol | |
| CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O | |
| 222.25 | |
| CHEBI:43253 | |
| 99+% |
| 28822-58-4 | |
| 100.0 | |
| White to Beige | |
| 99+% | |
| C10H14N4O2 | |
| 5 g | |
| 3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine | |
| APIXJSLKIYYUKG-UHFFFAOYSA-N | |
| 1-methyl-3-(2-methylpropyl)-7H-purine-2,6-dione | |
| 3758 | |
| 222.25 | |
| Fluffy Crystalline Powder |
Chemical Identifiers
| 28822-58-4 | |
| 222.25 | |
| APIXJSLKIYYUKG-UHFFFAOYSA-N | |
| 3758 | |
| CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O |
| C10H14N4O2 | |
| MFCD00005584 | |
| 3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine | |
| CHEBI:43253 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 249-259-3
RUO – Research Use Only