missing translation for 'onlineSavingsMsg'
Learn More

3-Iodothiophene 95.0+%, TCI America™

Catalog No. i03801g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03801G 1 g
I03805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03801G Supplier TCI America Supplier No. I03801G
Only null left

Chemical Identifiers

CAS 10486-61-0
Molecular Formula C4H3IS
Molecular Weight (g/mol) 210.03
MDL Number MFCD00037901
InChI Key WGKRMQIQXMJVFZ-UHFFFAOYSA-N
Synonym thiophene, 3-iodo, 3-iodothiophene, stab. with copper, 3-lodothiophene, 3-iodo-thiophene, 3-iodanylthiophene, 3-thienyl iodide, thien-3-yl iodide, pubchem16213, acmc-2098dn
PubChem CID 66332
IUPAC Name 3-iodothiophene
SMILES IC1=CSC=C1

Specifications

Melting Point -13°C
Color Yellow
Boiling Point 75°C
Quantity 1 g
Formula Weight 210.03
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Iodothiophene