missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopyrazolo[1,5-a]pyridine, 97%

Catalog No. AAH3525503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3525503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3525503 Supplier Thermo Scientific Chemicals Supplier No. H3525503
Only null left

CAS: 19363-99-6 | C7H5IN2 | 244.035 g/mol

Mostly used in the suzuki coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mostly used in the suzuki coupling reaction.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 19363-99-6
Molecular Formula C7H5IN2
Molecular Weight (g/mol) 244.035
MDL Number MFCD20483547
InChI Key PNNARKUCAKULAL-UHFFFAOYSA-N
Synonym 3-iodopyrazolo 1,5-a pyridine, 3-iodo-pyrazolo 1,5-a pyridine
PubChem CID 60170807
IUPAC Name 3-iodopyrazolo[1,5-a]pyridine
SMILES C1=CC2=C(C=NN2C=C1)I

Specifications

Melting Point 67°C to 69°C
Color Brown
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 244.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Iodopyrazolo[1,5-a]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only