missing translation for 'onlineSavingsMsg'
Learn More

3-Iodooxetane, 95%

Catalog No. AAH57302MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57302MD 250 mg
AAH57302ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57302MD Supplier Thermo Scientific Chemicals Supplier No. H57302MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 26272-85-5 | C3H5IO | 183.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26272-85-5
Molecular Formula C3H5IO
Molecular Weight (g/mol) 183.98
MDL Number MFCD08544403
InChI Key KBEIFKMKVCDETC-UHFFFAOYSA-N
Synonym 3-iodo-oxetane, 1,3-epoxy-2-iodopropane, oxetane, 3-iodo, 3-iodanyloxetane, 3-iodooxetane, acmc-1cgai, 2-iodo-1,3-epoxypropane, 1,3-?epoxy-?2-?iodopropane
PubChem CID 13356686
IUPAC Name 3-iodooxetane
SMILES IC1COC1

Specifications

Density 2.14
Boiling Point 166°C
Flash Point 54°C (129°F)
Quantity 250 mg
UN Number UN1993
Sensitivity Light sensitive
Formula Weight 183.98
Percent Purity 95%
Chemical Name or Material 3-Iodooxetane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only