missing translation for 'onlineSavingsMsg'
Learn More

3-Iodochromone, 97%

Catalog No. p-7032720
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34295MD 250 mg
AAH3429503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34295MD Supplier Thermo Scientific Chemicals Supplier No. H34295MD
Only null left

CAS: 122775-34-2 | C9H5IO2 | 272.04 g/mol

3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agent, light.

Chemical Identifiers

CAS 122775-34-2
Molecular Formula C9H5IO2
Molecular Weight (g/mol) 272.04
MDL Number MFCD09475514
InChI Key SZDBDDKQJHHRGH-UHFFFAOYSA-N
Synonym 3-iodochromone, 3-iodo-4h-chromen-4-one, 3-iodo-4h-benzopyran-4-one
PubChem CID 10901720
IUPAC Name 3-iodo-4H-chromen-4-one
SMILES IC1=COC2=CC=CC=C2C1=O

Specifications

Melting Point 95°C to 97°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 272.04
Percent Purity 97%
Chemical Name or Material 3-Iodochromone
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only