missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoanisole, 97%, Thermo Scientific™

Catalog No. p-3758224
Change view
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222020050 5g Glass bottle
AC222020250 25g Glass bottle
AC222021000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC222020050 Supplier Thermo Scientific Chemicals Supplier No. 222020050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 766-85-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.04
MDL Number MFCD00001048
InChI Key RSHBAGGASAJQCH-UHFFFAOYSA-N
Synonym 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole
PubChem CID 69839
IUPAC Name 1-iodo-3-methoxybenzene
SMILES COC1=CC(=CC=C1)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.9600g/mL
Boiling Point 244°C to 245°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6130 to 1.615
Linear Formula IC6H4OCH3
Quantity 5g
Beilstein 06, 208
Specific Gravity 1.96
Formula Weight 234.04
Percent Purity 97%
Chemical Name or Material 3-Iodoanisole
Show More Show Less
EINECSNumber 212-173-1
Recommended Storage Product may on darken on storage

RUO – Research Use Only