missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-9-phenylcarbazole 98.0+%, TCI America™
$67.11 - $93.21
Chemical Identifiers
| CAS | 502161-03-7 |
|---|---|
| Molecular Formula | C18H12IN |
| Molecular Weight (g/mol) | 369.205 |
| MDL Number | MFCD09261276 |
| InChI Key | PJUAIXDOXUXBDR-UHFFFAOYSA-N |
| Synonym | 3-iodo-n-phenylcarbazole, 3-iodo-9-phenyl-9h-carbazole, 9h-carbazole, 3-iodo-9-phenyl, 3-iodo-9-phenyl-carbazole, 3-iodo-n-phenyl-carbazole, ksc493m9j, 3-iodo-9-phenyl-9h-carbazol |
| PubChem CID | 11810740 |
| IUPAC Name | 3-iodo-9-phenylcarbazole |
| SMILES | C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=CC=CC=C42 |
Chemical Identifiers
| 502161-03-7 | |
| 369.205 | |
| PJUAIXDOXUXBDR-UHFFFAOYSA-N | |
| 11810740 | |
| C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=CC=CC=C42 |
| C18H12IN | |
| MFCD09261276 | |
| 3-iodo-n-phenylcarbazole, 3-iodo-9-phenyl-9h-carbazole, 9h-carbazole, 3-iodo-9-phenyl, 3-iodo-9-phenyl-carbazole, 3-iodo-n-phenyl-carbazole, ksc493m9j, 3-iodo-9-phenyl-9h-carbazol | |
| 3-iodo-9-phenylcarbazole |
Specifications
| 502161-03-7 | |
| White-Yellow | |
| MFCD09261276 | |
| 3-iodo-n-phenylcarbazole, 3-iodo-9-phenyl-9h-carbazole, 9h-carbazole, 3-iodo-9-phenyl, 3-iodo-9-phenyl-carbazole, 3-iodo-n-phenyl-carbazole, ksc493m9j, 3-iodo-9-phenyl-9h-carbazol | |
| C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=CC=CC=C42 | |
| 369.205 | |
| 369.21 | |
| Crystalline Powder |
| 106°C | |
| C18H12IN | |
| 1 g | |
| PJUAIXDOXUXBDR-UHFFFAOYSA-N | |
| 3-iodo-9-phenylcarbazole | |
| 11810740 | |
| ≥98.0% (GC) | |
| 3-Iodo-9-phenylcarbazole |
Safety and Handling
TSCA : No