missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1-methyl-1H-pyrazole, ≥97%, Thermo Scientific™

$1,006.08 - $1,006.08

Chemical Identifiers

CAS 92525-10-5
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.002
MDL Number MFCD08435890
InChI Key SITJXRWLFQGWCB-UHFFFAOYSA-N
PubChem CID 13334057
IUPAC Name 3-iodo-1-methylpyrazole
SMILES CN1C=CC(=N1)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC48311DA
SDS View Documents
Thermo Scientific Chemicals
CC48311DA
1 g Amber Glass Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

92525-10-5
208.002
SITJXRWLFQGWCB-UHFFFAOYSA-N
3-iodo-1-methylpyrazole
C4H5IN2
MFCD08435890
13334057
CN1C=CC(=N1)I

Specifications

92525-10-5
100.0
Amber Glass Bottle
MFCD08435890
3-iodo-1-methyl-1h-pyrazole, 1-methyl-3-iodopyrazole, 1h-pyrazole, 3-iodo-1-methyl, 3-iodo-1-methy-1h-pyrazole, iodo-n-methylpyrazole, 3-idio-1-methylpyrazole, 3-iodanyl-1-methyl-pyrazole, 1h-pyrazole,3-iodo-1-methyl, 3 5-iodopyrazole, 1-me
CN1C=CC(=N1)I
208.002
208
3-Iodo-1-methyl-1H-pyrazole
96.5
97%
C4H5IN2
1 g
SITJXRWLFQGWCB-UHFFFAOYSA-N
3-iodo-1-methylpyrazole
13334057
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'