missing translation for 'onlineSavingsMsg'
Learn More

3-Indolylacetonitrile, 97%

Catalog No. AC156650250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156650250 25 g Glass bottle
AC156650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156650250 Supplier Thermo Scientific Chemicals Supplier No. 156650250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 771-51-7 | C10H8N2 | 156.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 771-51-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00005628
InChI Key DMCPFOBLJMLSNX-UHFFFAOYSA-N
Synonym 3-indoleacetonitrile, indole-3-acetonitrile, 3-indolylacetonitrile, 2-1h-indol-3-yl acetonitrile, 1h-indole-3-acetonitrile, indolylacetonitrile, 3-cyanomethyl indole, indoleacetonitrile, indolylacetonitril, 3-indolacetonitrile
PubChem CID 351795
ChEBI CHEBI:17566
IUPAC Name 2-(1H-indol-3-yl)acetonitrile
SMILES C1=CC=C2C(=C1)C(=CN2)CC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 35°C to 37°C
Color Colorless to Yellow
Boiling Point 156°C to 160°C (0.2 mmHg)
Flash Point 206°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.6085 to 1.6105
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and toluene
Formula Weight 156.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Indolylacetonitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-232-1
RTECSNumber AM0700000
TSCA TSCA

RUO – Research Use Only