missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Indolepropanol 98.0+%, TCI America™
$195.73 - $583.44
Chemical Identifiers
| CAS | 3569-21-9 |
|---|---|
| Molecular Formula | C11H13NO |
| Molecular Weight (g/mol) | 175.23 |
| MDL Number | MFCD00013437 |
| InChI Key | LYPSVQXMCZIRGP-UHFFFAOYSA-N |
| Synonym | 3-(3-Hydroxypropyl)indole, Homotryptophol |
| PubChem CID | 77115 |
| IUPAC Name | 3-(1H-indol-3-yl)propan-1-ol |
| SMILES | OCCCC1=CNC2=CC=CC=C12 |
Chemical Identifiers
| 3569-21-9 | |
| 175.23 | |
| LYPSVQXMCZIRGP-UHFFFAOYSA-N | |
| 77115 | |
| OCCCC1=CNC2=CC=CC=C12 |
| C11H13NO | |
| MFCD00013437 | |
| 3-(3-Hydroxypropyl)indole, Homotryptophol | |
| 3-(1H-indol-3-yl)propan-1-ol |
Specifications
| 3569-21-9 | |
| 200°C | |
| MFCD00013437 | |
| 3-(3-Hydroxypropyl)indole, Homotryptophol | |
| OCCCC1=CNC2=CC=CC=C12 | |
| 175.23 | |
| 175.23 | |
| Liquid |
| Yellow | |
| C11H13NO | |
| 1 g | |
| LYPSVQXMCZIRGP-UHFFFAOYSA-N | |
| 3-(1H-indol-3-yl)propan-1-ol | |
| 77115 | |
| ≥98.0% (GC) | |
| 3-Indolepropanol |
Safety and Handling
TSCA : No