missing translation for 'onlineSavingsMsg'
Learn More

3-Indoleethanol 98.0+%, TCI America™

Catalog No. I00305G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00305G 5 g
I00301G 1 g
I003025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I00305G Supplier TCI America Supplier No. I00305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 526-55-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005659
InChI Key MBBOMCVGYCRMEA-UHFFFAOYSA-N
Synonym tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
PubChem CID 10685
ChEBI CHEBI:17890
IUPAC Name 2-(1H-indol-3-yl)ethan-1-ol
SMILES OCCC1=CNC2=CC=CC=C12

Specifications

Melting Point 59°C
Boiling Point 179°C
Quantity 5 g
Formula Weight 161.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indoleethanol
EINECSNumber (5)-5571
RTECSNumber KL3685000
TSCA Yes