missing translation for 'onlineSavingsMsg'
Learn More

3-Indolebutyric Acid 98.0+%, TCI America™

Catalog No. p-7131164
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00265G 5 g
I002625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00265G Supplier TCI America Supplier No. I00265G
Only null left

Chemical Identifiers

CAS 133-32-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00005664
InChI Key JTEDVYBZBROSJT-UHFFFAOYSA-N
Synonym indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
PubChem CID 8617
ChEBI CHEBI:33070
IUPAC Name 4-(1H-indol-3-yl)butanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 203.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indolebutyric Acid
EINECSNumber (5)-3368&(9)-0297
RTECSNumber NL5250000
TSCA Yes