missing translation for 'onlineSavingsMsg'
Learn More

3-Indolebutyric acid, 98%

Catalog No. ac160690250
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC160690250 25 g Glass bottle
AC160690050 5 g Glass bottle
AC160691000 100 g Glass bottle
3 options

Catalog No. AC160690250

Supplier: Thermo Scientific Chemicals 160690250

Only null left

CAS: 133-32-4 | C12H13NO2 | 203.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 133-32-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.24
MDL Number MFCD00005664
InChI Key JTEDVYBZBROSJT-UHFFFAOYSA-N
Synonym indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
PubChem CID 8617
ChEBI CHEBI:33070
IUPAC Name 4-(1H-indol-3-yl)butanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 120.0°C to 125.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 22, II, 54
Merck Index 15, 5005
Solubility Information Solubility in water: 250mg/l (20°C). Other solubilities: soluble in ethanol,ether,acetone,benzene,,dimethyl sulfoxide and petroleum ether,insoluble in chloroform,5% in ethanol: clear,brownish solution
Formula Weight 203.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Indolebutyric acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
EINECSNumber 205-101-5

RUO – Research Use Only