missing translation for 'onlineSavingsMsg'
Learn More

3-Indoleacetonitrile 98.0+%, TCI America™

Catalog No. i002425g
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I002425G 25 g
I00241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I002425G Supplier TCI America Supplier No. I002425G
Only null left

Chemical Identifiers

CAS 771-51-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00005628
InChI Key DMCPFOBLJMLSNX-UHFFFAOYSA-N
Synonym 3-indoleacetonitrile, indole-3-acetonitrile, 3-indolylacetonitrile, 2-1h-indol-3-yl acetonitrile, 1h-indole-3-acetonitrile, indolylacetonitrile, 3-cyanomethyl indole, indoleacetonitrile, indolylacetonitril, 3-indolacetonitrile
PubChem CID 351795
ChEBI CHEBI:17566
IUPAC Name 2-(1H-indol-3-yl)acetonitrile
SMILES C1=CC=C2C(=C1)C(=CN2)CC#N

Specifications

Melting Point 37°C
Quantity 25 g
UN Number 3439
Formula Weight 156.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indoleacetonitrile
RTECSNumber AM0700000
TSCA Yes
Recommended Storage Refrigerator