missing translation for 'onlineSavingsMsg'
Learn More

3-Indazolinone, 97%

Catalog No. p-7044322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0586006 5 g
AAL0586014 25 g
AAL0586022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0586006 Supplier Thermo Scientific Chemicals Supplier No. L0586006
Only null left

CAS: 7364-25-2 | C7H6N2O | 134.138 g/mol

3-Indazolinone is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Indazolinone is used as catalytic agent, petrochemical additive.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7364-25-2
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00005685
InChI Key SWEICGMKXPNXNU-UHFFFAOYSA-N
Synonym 1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol
PubChem CID 81829
IUPAC Name 1,2-dihydroindazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NN2

Specifications

Melting Point 248°C to 252°C
Quantity 5 g
Beilstein 3733
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 3-Indazolinone
EINECSNumber 230-904-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only