missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxytyramine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 189.64 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Hydroxytyramine Hydrochloride |
Safety and Handling
| EINECSNumber | (3)-2870 |
| RTECSNumber | UX1092000 |
| TSCA | Yes |